Product Details

SNP ID
rs73593269
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:24428742 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCTTTCCAGGTCCCCTGGACAGGG[G/T]TAGGGGGATTACCTGGTCGAAGTAG
Phenotype
MIM: 612978
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CBLN3 PubMed Links

Gene Details

Gene
CBLN3
Gene Name
cerebellin 3 precursor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039771.2 Intron NP_001034860.1
Gene
KHNYN
Gene Name
KH and NYN domain containing
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001290256.1 Intron NP_001277185.1
NM_001290257.1 Intron NP_001277186.1
NM_015299.2 Intron NP_056114.1
XM_005267474.4 Intron XP_005267531.1
XM_011536590.2 Intron XP_011534892.1
XM_017021114.1 Intron XP_016876603.1
Gene
NYNRIN
Gene Name
NYN domain and retroviral integrase containing
There are no transcripts associated with this gene.

View Full Product Details