Product Details

SNP ID
rs3745472
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:49645323 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGTTGCAAAGCATCTGCCTGGGTCC[C/T]CTGACGCTTGGTTCTTCCCCCCTTC
Phenotype
MIM: 165090
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RRAS PubMed Links

Gene Details

Gene
RRAS
Gene Name
related RAS viral (r-ras) oncogene homolog
There are no transcripts associated with this gene.

Gene
SCAF1
Gene Name
SR-related CTD associated factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021228.2 Intron NP_067051.2
XM_005259122.4 Intron XP_005259179.1
XM_011527194.2 Intron XP_011525496.1
XM_017027083.1 Intron XP_016882572.1

View Full Product Details