Product Details
- SNP ID
-
rs116144810
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:10672879 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GATGTGGGGCCCAAACTTCAGGCCA[C/G]GGCTTACCAGCCCCTGGACCTTGTG
- Phenotype
-
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
C8orf74
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs80181543] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- C8orf74
- Gene Name
- chromosome 8 open reading frame 74
- Gene
- MIR4286
- Gene Name
- microRNA 4286
There are no transcripts associated with this gene.
View Full Product Details