Product Details

SNP ID
rs10210695
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:127853074 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATGTTCCAGAAGCATATGAACTTC[C/T]GAATTTAGAAGTGTCTCAGCTGTTT
Phenotype
MIM: 606017
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AMMECR1L PubMed Links

Gene Details

Gene
AMMECR1L
Gene Name
AMMECR1 like
There are no transcripts associated with this gene.

Gene
LOC107985803
Gene Name
uncharacterized LOC107985803
There are no transcripts associated with this gene.

Gene
POLR2D
Gene Name
RNA polymerase II subunit D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004805.3 160 Silent Mutation TCA,TCG S35S NP_004796.1

View Full Product Details