Product Details

SNP ID
rs75805997
Assay Type
Functionally tested
NCBI dbSNP Submissions
6
Location
Chr.1:32241695 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGATAGGAACTGCCTGGGCAGCTG[A/G]CGACGCCGTCTGGACAGCTGTTAAG
Phenotype
MIM: 153390
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FAM167B PubMed Links

Gene Details

Gene
FAM167B
Gene Name
family with sequence similarity 167 member B
There are no transcripts associated with this gene.

Gene
LCK
Gene Name
LCK proto-oncogene, Src family tyrosine kinase
There are no transcripts associated with this gene.

Gene
MTMR9LP
Gene Name
myotubularin related protein 9-like, pseudogene
There are no transcripts associated with this gene.

View Full Product Details