Product Details
- SNP ID
-
rs13382312
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:127848080 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TGGTGTTATGCCTGGGGATGTGGTT[G/T]CTCCGAGCAGCAGTGATGTTTGGAG
- Phenotype
-
MIM: 606017
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
POLR2D
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs371001164] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- POLR2D
- Gene Name
- RNA polymerase II subunit D
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_004805.3 |
511 |
UTR 3 |
|
|
NP_004796.1 |
View Full Product Details