Product Details

SNP ID
rs61741445
Assay Type
Functionally tested
NCBI dbSNP Submissions
18
Location
Chr.1:34988872 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCATAAGTCTGGAAAAAACCTAAT[C/G]TTTTTTTCTTTGAAGGTGGAGAATC
Phenotype
MIM: 613567
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
TMEM35B PubMed Links

Gene Details

Gene
TMEM35B
Gene Name
transmembrane protein 35B
There are no transcripts associated with this gene.

Gene
ZMYM6
Gene Name
zinc finger MYM-type containing 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007167.3 2438 Missense Mutation ACA,AGA T737R NP_009098.3

View Full Product Details