Product Details

SNP ID
rs34663170
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:33210492 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTGATGGGCCTGTGGAGTGCAGGC[C/T]GCCTCCAGCGCAGCAGCAGCAGGCC
Phenotype
MIM: 603159 MIM: 607959
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LRP3 PubMed Links

Gene Details

Gene
LRP3
Gene Name
LDL receptor related protein 3
There are no transcripts associated with this gene.

Gene
SLC7A10
Gene Name
solute carrier family 7 member 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019849.2 1668 Missense Mutation NP_062823.1
XM_006723284.2 1668 Intron XP_006723347.1
XM_011527119.1 1668 Missense Mutation XP_011525421.1
XM_011527120.1 1668 Missense Mutation XP_011525422.1

View Full Product Details