Product Details

SNP ID
rs6437807
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:108914265 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CACAATTTCTCTTTTCTGAATTCAC[A/G]TCTCTTTTAACCCAGCCCTTTCTAC
Phenotype
MIM: 605128
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GUCA1C PubMed Links

Gene Details

Gene
GUCA1C
Gene Name
guanylate cyclase activator 1C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005459.3 Intron NP_005450.3
XM_005247910.3 Intron XP_005247967.1
XM_011513334.2 Intron XP_011511636.1

View Full Product Details