Product Details

SNP ID
rs4430318
Assay Type
Functionally tested
NCBI dbSNP Submissions
30
Location
Chr.1:40985764 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTGCCCTGGCAAGATTCTGGGCTAA[C/T]TGGGTCATGTTTGGATCTTAGCTTC
Phenotype
MIM: 123860
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CTPS1 PubMed Links

Gene Details

Gene
CTPS1
Gene Name
CTP synthase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001301237.1 Intron NP_001288166.1
NM_001905.3 Intron NP_001896.2

View Full Product Details