Product Details

SNP ID
rs626089
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:10470991 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATATGTAGCTTTGTTAGATACTGT[G/A]AAATTCCCTTTTTTGGGGCTTATGG
Phenotype
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
BAGE2 PubMed Links

Gene Details

Gene
BAGE2
Gene Name
B melanoma antigen family member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_182482.2 Intron NP_872288.2
Gene
LOC105378260
Gene Name
histone-lysine N-methyltransferase 2C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_017028520.1 Intron XP_016884009.1
XM_017028521.1 Intron XP_016884010.1

View Full Product Details