Product Details

SNP ID
rs6503437
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:45440786 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCAGAGCCTGCACCCCTGTCCACCT[A/G]GCAAATGCCTAGAGATGCAGGCTGT
Phenotype
MIM: 610591 MIM: 611466
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ARHGAP27 PubMed Links

Gene Details

Gene
ARHGAP27
Gene Name
Rho GTPase activating protein 27
There are no transcripts associated with this gene.

Gene
PLEKHM1
Gene Name
pleckstrin homology and RUN domain containing M1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014798.2 Intron NP_055613.1
XM_006722201.3 Intron XP_006722264.1
XM_011525523.2 Intron XP_011523825.1
XM_011525524.1 Intron XP_011523826.1
XM_011525525.1 Intron XP_011523827.1
XM_011525526.1 Intron XP_011523828.1
XM_011525528.2 Intron XP_011523830.1
XM_017025451.1 Intron XP_016880940.1
XM_017025452.1 Intron XP_016880941.1
XM_017025453.1 Intron XP_016880942.1
XM_017025454.1 Intron XP_016880943.1

View Full Product Details