Product Details

SNP ID
rs12402335
Assay Type
Functionally tested
NCBI dbSNP Submissions
28
Location
Chr.1:114593474 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACTGTAACATCCAAAACTAAACTTT[A/T]TATTACTCCTCTAAATCTGCTCCTT
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
DENND2C PubMed Links

Gene Details

Gene
DENND2C
Gene Name
DENN domain containing 2C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256404.1 Intron NP_001243333.1
NM_198459.3 Intron NP_940861.3

View Full Product Details