Product Details

SNP ID
rs7480975
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:45099826 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTCTTATGCATTCTTTCTAGCTTA[A/G]ACTGCCAATTCTGAAGCTCATGCAC
Phenotype
MIM: 616347
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PRDM11 PubMed Links

Gene Details

Gene
PRDM11
Gene Name
PR domain 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256695.1 Intron NP_001243624.1
NM_001256696.1 Intron NP_001243625.1
XM_011520222.2 Intron XP_011518524.1
XM_011520223.2 Intron XP_011518525.1
XM_011520224.2 Intron XP_011518526.1
XM_011520225.2 Intron XP_011518527.1
XM_011520226.2 Intron XP_011518528.1
XM_011520227.2 Intron XP_011518529.1
XM_011520228.2 Intron XP_011518530.1
XM_011520230.2 Intron XP_011518532.1
XM_011520231.2 Intron XP_011518533.1
XM_017018024.1 Intron XP_016873513.1

View Full Product Details