Product Details
- SNP ID
-
rs10828268
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:18147718 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AATTAACTAAAACAATTACATATAT[A/T]TTTTTTTTCAAATTTAGTCAGCTAA
- Phenotype
-
MIM: 600003
- Polymorphism
- A/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
CACNB2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs531900535] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CACNB2
- Gene Name
- calcium voltage-gated channel auxiliary subunit beta 2
View Full Product Details