Product Details

SNP ID
rs441549
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:52543026 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAAAGGGGGCATCAGTGCAGCCCAG[A/G]TCCCCCCTGCTGCCTCGTGTGTGGC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF808 PubMed Links

Gene Details

Gene
ZNF808
Gene Name
zinc finger protein 808
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039886.3 Intron NP_001034975.2
NM_001321424.1 Intron NP_001308353.1
NM_001321425.1 Intron NP_001308354.1
XM_005258909.3 Intron XP_005258966.1

View Full Product Details