Product Details

SNP ID
rs1112851
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:13306540 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTATCCCAAACCCTACAAAAATAA[A/G]GAGATATAATCAAATTATATGAGTT
Phenotype
MIM: 612655
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TBC1D7 PubMed Links

Gene Details

Gene
TBC1D7
Gene Name
TBC1 domain family member 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001143964.3 Intron NP_001137436.1
NM_001143965.3 Intron NP_001137437.1
NM_001143966.3 Intron NP_001137438.1
NM_001258457.2 Intron NP_001245386.1
NM_001318805.1 Intron NP_001305734.1
NM_001318806.1 Intron NP_001305735.1
NM_016495.5 Intron NP_057579.1
Gene
TBC1D7-LOC100130357
Gene Name
TBC1D7-LOC100130357 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318809.1 Intron NP_001305738.1

View Full Product Details