Product Details

SNP ID
rs11879865
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:4970037 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTGCTTCCGCCGAGACCCCTTTTCT[A/G]TGTCCCCCGTGGACATCCCCCTGGG
Phenotype
MIM: 609765 MIM: 607990
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KDM4B PubMed Links

Gene Details

Gene
KDM4B
Gene Name
lysine demethylase 4B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015015.2 Intron NP_055830.1
XM_005259521.3 Intron XP_005259578.2
XM_011527814.2 Intron XP_011526116.1
XM_011527816.2 Intron XP_011526118.1
XM_011527817.2 Intron XP_011526119.1
XM_011527818.2 Intron XP_011526120.1
XM_011527819.1 Intron XP_011526121.1
XM_011527820.1 Intron XP_011526122.1
XM_011527821.1 Intron XP_011526123.1
XM_011527822.1 Intron XP_011526124.1
XM_017026503.1 Intron XP_016881992.1
XM_017026504.1 Intron XP_016881993.1
XM_017026505.1 Intron XP_016881994.1
Gene
UHRF1
Gene Name
ubiquitin like with PHD and ring finger domains 1
There are no transcripts associated with this gene.

View Full Product Details