Product Details

SNP ID
rs3747421
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:14009058 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACGCTTCATCTCGGCCTGGCGCCGC[A/T]CACCAGGCCTCTCAGTTGGGGCTTC
Phenotype
MIM: 300962
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
GEMIN8 PubMed Links

Gene Details

Gene
GEMIN8
Gene Name
gem nuclear organelle associated protein 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042479.1 927 Missense Mutation GAG,GTG E195V NP_001035944.1
NM_001042480.1 927 Missense Mutation GAG,GTG E195V NP_001035945.1
NM_017856.2 927 Missense Mutation GAG,GTG E195V NP_060326.1
XM_005274555.2 927 Missense Mutation GAG,GTG E296V XP_005274612.1
XM_017029617.1 927 Missense Mutation GAG,GTG E195V XP_016885106.1
XM_017029618.1 927 Missense Mutation GAG,GTG E195V XP_016885107.1

View Full Product Details