Product Details

SNP ID
rs3753261
Assay Type
Functionally tested
NCBI dbSNP Submissions
38
Location
Chr.1:119367866 on Build GRCh38
Set Membership
HapMap JSNP
Context Sequence [VIC/FAM]
TGGTCATCGGTACAGCTCAGGATTT[C/T]GGCCTCCTCTGCTCCGTGTGTTGGC
Phenotype
MIM: 605176
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HAO2 PubMed Links

Gene Details

Gene
HAO2
Gene Name
hydroxyacid oxidase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005783.2 Intron NP_001005783.2
NM_016527.3 Intron NP_057611.1
XM_011541558.1 Intron XP_011539860.1
XM_011541559.1 Intron XP_011539861.1
XM_011541560.1 Intron XP_011539862.1
XM_011541561.1 Intron XP_011539863.1
XM_017001421.1 Intron XP_016856910.1
Gene
HAO2-IT1
Gene Name
HAO2 intronic transcript 1
There are no transcripts associated with this gene.

View Full Product Details