Product Details

SNP ID
rs3790688
Assay Type
Functionally tested
NCBI dbSNP Submissions
65
Location
Chr.1:68047801 on Build GRCh38
Set Membership
HapMap JSNP
Context Sequence [VIC/FAM]
CAAGGTGACATTTAGAGTGACACCG[C/T]CGCCCTGGTTAAAGCTACAGAGTCA
Phenotype
MIM: 605193 MIM: 615406
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DIRAS3 PubMed Links

Gene Details

Gene
DIRAS3
Gene Name
DIRAS family GTPase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004675.3 Intron NP_004666.1
XM_017002773.1 Intron XP_016858262.1
Gene
GNG12-AS1
Gene Name
GNG12 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details