Product Details

SNP ID
rs3750135
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.7:156950085 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGGCCTGGGAGGCCGAAGCGGAGC[A/C]GAAGAAGCCAGCGGTCACCGGCAGG
Phenotype
MIM: 611269
Polymorphism
A/C, Transversion Substitution
Allele Nomenclature
Literature Links
NOM1 PubMed Links
Additional Information
For this assay, SNP(s) [rs147347140,rs376182769] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
NOM1
Gene Name
nucleolar protein with MIF4G domain 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138400.1 363 Silent Mutation GCA,GCC A116A NP_612409.1

View Full Product Details