Product Details

SNP ID
rs3750734
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:73119882 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TATAGCCACCTCAAACCCTTTTTGG[A/G]GCAAATTAGGTTGTAAATAAATAAA
Phenotype
MIM: 609233
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NUDT13 PubMed Links

Gene Details

Gene
NUDT13
Gene Name
nudix hydrolase 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001283014.1 Intron NP_001269943.1
NM_001283015.1 Intron NP_001269944.1
NM_001283016.1 Intron NP_001269945.1
NM_001283017.1 Intron NP_001269946.1
NM_001283019.1 Intron NP_001269948.1
NM_015901.5 Intron NP_056985.3
XM_017016033.1 Intron XP_016871522.1

View Full Product Details