Product Details

SNP ID
rs3795499
Assay Type
Functionally tested
NCBI dbSNP Submissions
25
Location
Chr.1:36341851 on Build GRCh38
Set Membership
HapMap JSNP
Context Sequence [VIC/FAM]
GCCGTCGCACCGGTGGTGCGCTGCC[A/G]AACTGCCGCTTGGGTACCCAGCTCC
Phenotype
MIM: 609437
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
STK40 PubMed Links

Gene Details

Gene
STK40
Gene Name
serine/threonine kinase 40
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282546.1 1662 Silent Mutation TTC,TTT F409F NP_001269475.1
NM_001282547.1 1662 Silent Mutation TTC,TTT F404F NP_001269476.1
NM_032017.2 1662 Silent Mutation TTC,TTT F404F NP_114406.1

View Full Product Details