Product Details

SNP ID
rs7654520
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:188139015 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTAGGCTTTGGTCAAGCAGAACAAA[A/C]GGGGCTAATGAGAAAAGGACTATGT
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
TRIML1 PubMed Links

Gene Details

Gene
TRIML1
Gene Name
tripartite motif family like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178556.3 Intron NP_848651.2
XM_006714207.3 Intron XP_006714270.1
XM_011531919.1 Intron XP_011530221.1

View Full Product Details