Product Details
- SNP ID
-
hCV27838391
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:50117279 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGCACTATCAGCAAAACACTCTATC[A/C]CTTTGAACGAAGGCATCCAAATGGC
- Phenotype
-
MIM: 601247
MIM: 615260
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
SOS2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs148504789] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- SOS2
- Gene Name
- SOS Ras/Rho guanine nucleotide exchange factor 2
There are no transcripts associated with this gene.
- Gene
- VCPKMT
- Gene Name
- valosin containing protein lysine methyltransferase
View Full Product Details