Product Details

SNP ID
rs147851913
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:55181590 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCATAGGACTCATCTGAAGCCCACC[A/T]GACCACTCTCTCCTGGGGAAACCAG
Phenotype
MIM: 602510 MIM: 600782
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
PTPRH PubMed Links

Gene Details

Gene
PTPRH
Gene Name
protein tyrosine phosphatase, receptor type H
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001161440.2 3606 UTR 3 NP_001154912.2
NM_002842.4 3606 UTR 3 NP_002833.4
XM_011527183.1 3606 UTR 3 XP_011525485.1
XM_011527188.1 3606 UTR 3 XP_011525490.1
XM_011527190.1 3606 Intron XP_011525492.1
XM_017027056.1 3606 UTR 3 XP_016882545.1
XM_017027057.1 3606 UTR 3 XP_016882546.1
XM_017027058.1 3606 UTR 3 XP_016882547.1
XM_017027059.1 3606 UTR 3 XP_016882548.1
XM_017027060.1 3606 UTR 3 XP_016882549.1
XM_017027061.1 3606 UTR 3 XP_016882550.1
XM_017027062.1 3606 UTR 3 XP_016882551.1
XM_017027063.1 3606 UTR 3 XP_016882552.1
XM_017027064.1 3606 Intron XP_016882553.1
Gene
SYT5
Gene Name
synaptotagmin 5
There are no transcripts associated with this gene.

View Full Product Details