Product Details

SNP ID
rs73444259
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:135240138 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGTAAAATTAACCCCTGAAATTAA[C/T]AAGTGTCTGGTGAGTATCTGCAATG
Phenotype
MIM: 609987
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
STRA8 PubMed Links

Gene Details

Gene
STRA8
Gene Name
stimulated by retinoic acid 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_182489.1 320 Intron NP_872295.1
XM_011516137.2 320 UTR 5 XP_011514439.1
XM_011516138.2 320 Intron XP_011514440.1
XM_011516139.2 320 UTR 5 XP_011514441.1
XM_017012150.1 320 UTR 5 XP_016867639.1

View Full Product Details