Product Details

SNP ID
hCV27862934
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:48688056 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAAAAAAAGAGATAGGTGCCATGC[A/G]GGTGAAATCGGTGAAGATGAGGATT
Phenotype
MIM: 605358 MIM: 152790
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GTF2A1L PubMed Links

Gene Details

Gene
GTF2A1L
Gene Name
general transcription factor IIA subunit 1 like
There are no transcripts associated with this gene.

Gene
LHCGR
Gene Name
luteinizing hormone/choriogonadotropin receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000233.3 1811 Missense Mutation CGC,TGC R581C NP_000224.2
Gene
STON1-GTF2A1L
Gene Name
STON1-GTF2A1L readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001198593.1 1811 Intron NP_001185522.1
NM_001198594.1 1811 Intron NP_001185523.1
NM_172311.2 1811 Intron NP_758515.1

View Full Product Details