Product Details

SNP ID
rs4653123
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:35185558 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGTGCCTAAATAGCAGACCCCTCA[C/G]GTATGCAAACACAGCAGCCTACACT
Phenotype
MIM: 605199
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
SFPQ PubMed Links

Gene Details

Gene
SFPQ
Gene Name
splicing factor proline and glutamine rich
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005066.2 Intron NP_005057.1

View Full Product Details