Product Details

SNP ID
rs4898526
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:49632759 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTCTGGGATATTGATGGGACCAGG[C/G]GACTATTTACTGGGGCTGGAATACG
Phenotype
MIM: 616288
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
FMNL3 PubMed Links

Gene Details

Gene
FMNL3
Gene Name
formin like 3
There are no transcripts associated with this gene.

Gene
PRPF40B
Gene Name
pre-mRNA processing factor 40 homolog B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001031698.2 Intron NP_001026868.2
NM_012272.2 Intron NP_036404.1

View Full Product Details