Product Details

SNP ID
rs6955913
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:82754059 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAAGGCTGAGGCAACAGCCTTTATA[C/T]GGCACAGTTTACATAAGTTATCAAG
Phenotype
MIM: 604918
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PCLO PubMed Links

Gene Details

Gene
PCLO
Gene Name
piccolo presynaptic cytomatrix protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014510.2 20234 Intron NP_055325.2
NM_033026.5 20234 UTR 3 NP_149015.2
XM_017012006.1 20234 Intron XP_016867495.1
XM_017012007.1 20234 Intron XP_016867496.1

View Full Product Details