Product Details

SNP ID
rs4900988
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:50105043 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AACTCAAAAGGTAGGCTTTTTTTTT[A/T]AATCTAATTTCGTTCACCTTTAAAA
Phenotype
MIM: 615260
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
LINC01599 PubMed Links

Gene Details

Gene
LINC01599
Gene Name
long intergenic non-protein coding RNA 1599
There are no transcripts associated with this gene.

Gene
VCPKMT
Gene Name
valosin containing protein lysine methyltransferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001040662.1 2267 Intron NP_001035752.1
NM_024558.2 2267 Intron NP_078834.2
XM_005268060.3 2267 Intron XP_005268117.1
XM_017021640.1 2267 UTR 3 XP_016877129.1
XM_017021641.1 2267 UTR 3 XP_016877130.1
XM_017021642.1 2267 Intron XP_016877131.1

View Full Product Details