Product Details

SNP ID
rs7036967
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:89324869 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGTTTAGTCTAGCAGCTTGGGATAA[A/C]TTGAGGCAATAGCAACCTTCAAAGC
Phenotype
MIM: 116901 MIM: 607693
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CKS2 PubMed Links

Gene Details

Gene
CKS2
Gene Name
CDC28 protein kinase regulatory subunit 2
There are no transcripts associated with this gene.

Gene
SECISBP2
Gene Name
SECIS binding protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282688.1 Intron NP_001269617.1
NM_001282689.1 Intron NP_001269618.1
NM_001282690.1 Intron NP_001269619.1
NM_024077.4 Intron NP_076982.3
XM_005252196.2 Intron XP_005252253.1
XM_005252202.3 Intron XP_005252259.1
XM_006717282.2 Intron XP_006717345.1
XM_011519000.2 Intron XP_011517302.1
XM_011519001.1 Intron XP_011517303.1
XM_011519002.1 Intron XP_011517304.1
XM_011519003.1 Intron XP_011517305.1
XM_017015122.1 Intron XP_016870611.1
XM_017015123.1 Intron XP_016870612.1
XM_017015124.1 Intron XP_016870613.1
XM_017015125.1 Intron XP_016870614.1

View Full Product Details