Product Details
- SNP ID
-
rs6128440
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:58825649 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AACCGTGAAGGTGGTGCGAAAATAA[C/T]TGAAGTCAGCAAAAATGACCTCGAT
- Phenotype
-
MIM: 610540
MIM: 610945
MIM: 614914
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
GNAS-AS1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs76955929] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- GNAS-AS1
- Gene Name
- GNAS antisense RNA 1
There are no transcripts associated with this gene.
- Gene
- MIR296
- Gene Name
- microRNA 296
There are no transcripts associated with this gene.
- Gene
- MIR298
- Gene Name
- microRNA 298
There are no transcripts associated with this gene.
View Full Product Details