Product Details
- SNP ID
-
rs6032452
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:45708881 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GGTATCCATAATTATACAAGCATCA[A/G]GCTCCACAAAGCCTGGGCCTTTCTC
- Phenotype
-
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
MIR3617
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs75748348] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- MIR3617
- Gene Name
- microRNA 3617
There are no transcripts associated with this gene.
- Gene
- WFDC10B
- Gene Name
- WAP four-disulfide core domain 10B
- Gene
- WFDC13
- Gene Name
- WAP four-disulfide core domain 13
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_172005.1 |
19 |
Intron |
|
|
NP_742002.1 |
- Gene
- WFDC3
- Gene Name
- WAP four-disulfide core domain 3
View Full Product Details