Product Details

SNP ID
rs10473688
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:24490596 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAAATGCTATGATAAAAGTAGAAAA[A/G]TGGAAACATCTATAGAATGAAGTAG
Phenotype
MIM: 604555
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CDH10 PubMed Links

Gene Details

Gene
CDH10
Gene Name
cadherin 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317222.1 Intron NP_001304151.1
NM_001317224.1 Intron NP_001304153.1
NM_006727.4 Intron NP_006718.2
XM_011513923.2 Intron XP_011512225.1
XM_017008915.1 Intron XP_016864404.1

View Full Product Details