Product Details

SNP ID
rs7278427
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:10417840 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTTTTTTCTTAAATCCCATCAACC[G/A]TGACTGGTATCCATCCATGTCCTAG
Phenotype
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
BAGE2 PubMed Links

Gene Details

Gene
BAGE2
Gene Name
B melanoma antigen family member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_182482.2 Intron NP_872288.2

View Full Product Details