Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_000233.3 | 2256 | UTR 3 | NP_000224.2 | ||
XM_005264309.3 | 2256 | UTR 3 | XP_005264366.1 | ||
XM_006712015.3 | 2256 | UTR 3 | XP_006712078.1 | ||
XM_011532828.1 | 2256 | UTR 3 | XP_011531130.1 | ||
XM_011532831.1 | 2256 | UTR 3 | XP_011531133.1 | ||
XM_011532834.2 | 2256 | UTR 3 | XP_011531136.1 | ||
XM_017004089.1 | 2256 | UTR 3 | XP_016859578.1 | ||
XM_017004090.1 | 2256 | UTR 3 | XP_016859579.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001198593.1 | 2256 | Intron | NP_001185522.1 | ||
NM_001198594.1 | 2256 | Intron | NP_001185523.1 | ||
NM_172311.2 | 2256 | Intron | NP_758515.1 |