Product Details

SNP ID
rs10159583
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:121993154 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGCAATGTTGCAAGACCCCTGCCTC[A/T]ACAACAAACAAACAAAACAAAACTC
Phenotype
MIM: 605302
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
TACC2 PubMed Links

Gene Details

Gene
TACC2
Gene Name
transforming acidic coiled-coil containing protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001291876.1 Intron NP_001278805.1
NM_001291877.1 Intron NP_001278806.1
NM_001291878.1 Intron NP_001278807.1
NM_001291879.1 Intron NP_001278808.1
NM_006997.3 Intron NP_008928.1
NM_206860.2 Intron NP_996742.1
NM_206861.2 Intron NP_996743.1
NM_206862.3 Intron NP_996744.3
XM_005269388.1 Intron XP_005269445.1
XM_005269389.4 Intron XP_005269446.1
XM_005269390.1 Intron XP_005269447.1
XM_005269391.1 Intron XP_005269448.1
XM_005269392.1 Intron XP_005269449.1
XM_005269393.1 Intron XP_005269450.1
XM_005269394.1 Intron XP_005269451.1
XM_005269395.1 Intron XP_005269452.1
XM_005269396.1 Intron XP_005269453.1
XM_005269397.1 Intron XP_005269454.1
XM_005269399.1 Intron XP_005269456.1
XM_006717548.1 Intron XP_006717611.1
XM_006717552.1 Intron XP_006717615.1
XM_006717553.1 Intron XP_006717616.1
XM_006717554.1 Intron XP_006717617.1
XM_011539126.1 Intron XP_011537428.1
XM_011539128.1 Intron XP_011537430.1
XM_011539129.1 Intron XP_011537431.1
XM_011539130.1 Intron XP_011537432.1
XM_011539131.1 Intron XP_011537433.1
XM_011539132.1 Intron XP_011537434.1
XM_011539133.1 Intron XP_011537435.1
XM_011539134.1 Intron XP_011537436.1
XM_017015484.1 Intron XP_016870973.1
XM_017015485.1 Intron XP_016870974.1
XM_017015486.1 Intron XP_016870975.1
XM_017015487.1 Intron XP_016870976.1
XM_017015488.1 Intron XP_016870977.1
XM_017015489.1 Intron XP_016870978.1
XM_017015490.1 Intron XP_016870979.1
XM_017015491.1 Intron XP_016870980.1
XM_017015492.1 Intron XP_016870981.1
XM_017015493.1 Intron XP_016870982.1
XM_017015494.1 Intron XP_016870983.1
XM_017015495.1 Intron XP_016870984.1
XM_017015496.1 Intron XP_016870985.1
XM_017015497.1 Intron XP_016870986.1
XM_017015498.1 Intron XP_016870987.1
XM_017015499.1 Intron XP_016870988.1

View Full Product Details