Product Details

SNP ID
hCV30503092
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:169862558 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTGGTAGCATATCTTATTGAAATG[C/T]GGCTTCTTAAGTATGACAAGTGTAT
Phenotype
MIM: 603122
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DOCK2 PubMed Links

Gene Details

Gene
DOCK2
Gene Name
dedicator of cytokinesis 2
There are no transcripts associated with this gene.

Gene
FAM196B
Gene Name
family with sequence similarity 196 member B
There are no transcripts associated with this gene.

View Full Product Details