Product Details
- SNP ID
-
rs7518470
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
35
- Location
-
Chr.1:62198115 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ATGTCCCCATACCCAAGGAGAGAGA[C/T]GTACTAACTAGAAGTCGAAGTTGGC
- Phenotype
-
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
L1TD1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs145693231] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- L1TD1
- Gene Name
- LINE1 type transposase domain containing 1
There are no transcripts associated with this gene.
View Full Product Details