Product Details

SNP ID
rs11989080
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:69672040 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAGAAAGCTGTCATTTTCTTTCCCT[A/G]TTATGCATTTCAAATTACAGTTTCT
Phenotype
MIM: 613543
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SLCO5A1 PubMed Links

Gene Details

Gene
SLCO5A1
Gene Name
solute carrier organic anion transporter family member 5A1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001146008.1 3918 Intron NP_001139480.1
NM_001146009.1 3918 Intron NP_001139481.1
NM_030958.2 3918 Intron NP_112220.2
XM_005251313.2 3918 UTR 3 XP_005251370.1
XM_017013883.1 3918 UTR 3 XP_016869372.1
XM_017013884.1 3918 Intron XP_016869373.1
XM_017013885.1 3918 UTR 3 XP_016869374.1
XM_017013886.1 3918 Intron XP_016869375.1

View Full Product Details