Product Details

SNP ID
rs11202498
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:87668490 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCATTTGCCTTTTCTAATTGAAG[C/G]ACTCCACCCGCACCTTTATATTATC
Phenotype
MIM: 603005
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
PAPSS2 PubMed Links

Gene Details

Gene
PAPSS2
Gene Name
3'-phosphoadenosine 5'-phosphosulfate synthase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001015880.1 Intron NP_001015880.1
NM_004670.3 Intron NP_004661.2

View Full Product Details