Product Details

SNP ID
rs10749750
Assay Type
Functionally tested
NCBI dbSNP Submissions
67
Location
Chr.1:65154045 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CACTGGAAAGCCAAGGAAGGGTTTT[A/C]CATAGGACAATGACATGAAAAGATT
Phenotype
MIM: 103030
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
AK4 PubMed Links

Gene Details

Gene
AK4
Gene Name
adenylate kinase 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005353.2 Intron NP_001005353.1
NM_013410.3 Intron NP_037542.1
NM_203464.2 Intron NP_982289.1
XM_017000612.1 Intron XP_016856101.1
XM_017000613.1 Intron XP_016856102.1

View Full Product Details