Product Details

SNP ID
rs10985254
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:121462804 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGCAGTTAAGTGCAATACTTTGTT[A/C]TGAGCTTTCTGACAACCAACCCAAC
Phenotype
MIM: 104175
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
GGTA1P PubMed Links

Gene Details

Gene
GGTA1P
Gene Name
glycoprotein, alpha-galactosyltransferase 1 pseudogene
There are no transcripts associated with this gene.

View Full Product Details