Product Details

SNP ID
hCV31023070
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:86278848 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAACATCTTTTTATTCCTGCCATTG[G/T]GATTTAGGTATTAAAGATTAAATAA
Phenotype
MIM: 611006 MIM: 613467
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ISCA1 PubMed Links

Gene Details

Gene
ISCA1
Gene Name
iron-sulfur cluster assembly 1
There are no transcripts associated with this gene.

Gene
ZCCHC6
Gene Name
zinc finger CCHC-type containing 6
There are no transcripts associated with this gene.

View Full Product Details