Product Details

SNP ID
rs13381300
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:2572096 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATGAGAGCTACATTCTAAGTGGAGA[G/T]TAAACGTGGACACACAGGTGTGAGT
Phenotype
MIM: 607272
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
METTL4 PubMed Links

Gene Details

Gene
METTL4
Gene Name
methyltransferase like 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308401.1 Intron NP_001295330.1
NM_022840.4 Intron NP_073751.3
XM_005258132.3 Intron XP_005258189.1
XM_005258133.2 Intron XP_005258190.1
XM_011525730.2 Intron XP_011524032.1
Gene
NDC80
Gene Name
NDC80, kinetochore complex component
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006101.2 Intron NP_006092.1

View Full Product Details