Product Details

SNP ID
rs13031596
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:86447494 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGCTGAATAGATATTAGATAATTG[G/T]GTTGAAGATCCTCTTTCCCCTGCTC
Phenotype
MIM: 611512
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
KDM3A PubMed Links

Gene Details

Gene
KDM3A
Gene Name
lysine demethylase 3A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001146688.1 Intron NP_001140160.1
NM_018433.5 Intron NP_060903.2
XM_006712051.3 Intron XP_006712114.1
XM_017004493.1 Intron XP_016859982.1
XM_017004494.1 Intron XP_016859983.1

View Full Product Details