Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001122838.1 | 2832 | UTR 3 | NP_001116310.1 | ||
NM_198990.4 | 2832 | UTR 3 | NP_945341.3 | ||
XM_005250214.4 | 2832 | UTR 3 | XP_005250271.2 | ||
XM_005250215.2 | 2832 | UTR 3 | XP_005250272.1 | ||
XM_005250218.2 | 2832 | UTR 3 | XP_005250275.1 | ||
XM_011515933.2 | 2832 | UTR 3 | XP_011514235.1 | ||
XM_011515934.2 | 2832 | UTR 3 | XP_011514236.1 | ||
XM_017011854.1 | 2832 | UTR 3 | XP_016867343.1 | ||
XM_017011855.1 | 2832 | UTR 3 | XP_016867344.1 | ||
XM_017011856.1 | 2832 | Intron | XP_016867345.1 | ||
XM_017011857.1 | 2832 | Intron | XP_016867346.1 |